Assay Details
| Gene Symbol: AT5G53770 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124756.4 | NP_568798.1 | 4 - 5 | 708 | 106 | |
| GenBank mRNA | BT002764.1 | - | 4 - 5 | 631 | 106 | |
More Information
Set Membership: |
|

AT5G53770
Genomic Map