Assay Details
| Gene Symbol: AT5G53770 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124756.4 | NP_568798.1 | 6 - 7 | 826 | 139 | |
| GenBank mRNA | BT002764.1 | - | 6 - 7 | 749 | 139 | |
More Information
Set Membership: |
|

AT5G53770
Genomic Map