Assay Details
| Gene Symbol: AT5G53930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124772.3 | NP_200204.1 | 2 - 3 | 1669 | 84 | |
| GenBank mRNA | AK119009.1 | - | 2 - 3 | 1662 | 84 | |
| BT005486.1 | - | 2 - 3 | 1518 | 84 | ||
More Information
Set Membership: |
|

AT5G53930
Genomic Map