Assay Details
| Gene Symbol: TAT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124776.5 | NP_200208.1 | 4 - 5 | 1070 | 109 | |
| GenBank mRNA | BT001912.1 | - | 4 - 5 | 865 | 109 | |
More Information
Set Membership: |
|

TAT7
Genomic Map