Assay Details
| Gene Symbol: PORA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036995.1 | NP_001032072.1 | 1 - 2 | 94 | 87 | |
| NM_124799.4 | NP_200230.1 | 1 - 2 | 96 | 87 | ||
| GenBank mRNA | AK317329.1 | - | 1 - 2 | 95 | 87 | |
| BT003853.1 | - | 1 - 2 | 93 | 87 | ||
| BT005080.1 | - | 1 - 2 | 55 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001036995.1 transcripts.
Set Membership: |
|

PORA
Genomic Map