Assay Details
| Gene Symbol: MOR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336567.1 | NP_001325149.1 | 28 - 29 | 3472 | 82 | |
| NM_129117.5 | NP_565811.2 | 28 - 29 | 3166 | 82 | ||
| GenBank mRNA | AF367246.1 | - | 28 - 29 | 2912 | 82 | |
More Information
Set Membership: |
|

MOR1
Genomic Map