Assay Details
Gene Symbol: AT2G35760 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129134.4 | NP_565822.1 | 2 - 3 | 738 | 97 | |
GenBank mRNA | AF462840.1 | - | 2 - 3 | 497 | 97 | |
AY124824.1 | - | 2 - 3 | 358 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
