Assay Details
Gene Symbol: ABI5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336590.1 | NP_001324735.1 | 2 - 3 | 1294 | 92 | |
NM_001336591.1 | NP_001324734.1 | 3 - 4 | 1488 | 186 | ||
NM_129185.4 | NP_565840.1 | 3 - 4 | 1434 | 92 | ||
GenBank mRNA | AF334206.1 | - | 3 - 4 | 1352 | 92 | |
BT026517.1 | - | 2 - 3 | 1205 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
