Assay Details
Gene Symbol: ABI5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336590.1 | NP_001324735.1 | 3 - 4 | 1314 | 65 | |
NM_129185.4 | NP_565840.1 | 4 - 5 | 1454 | 65 | ||
GenBank mRNA | AF334206.1 | - | 4 - 5 | 1372 | 65 | |
BT026517.1 | - | 3 - 4 | 1225 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
