Assay Details
| Gene Symbol: FBA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129203.3 | NP_181187.1 | 2 - 3 | 624 | 94 | |
| GenBank mRNA | AK226842.1 | - | 324 | 94 | ||
| AY034897.1 | - | 2 - 3 | 323 | 94 | ||
| AY063044.1 | - | 2 - 3 | 257 | 94 | ||
| AY085114.1 | - | 2 - 3 | 320 | 94 | ||
More Information
Set Membership: |
|

FBA6
Genomic Map