Assay Details
| Gene Symbol: DML1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129207.5 | NP_181190.3 | 15 - 16 | 3942 | 89 | |
| GenBank mRNA | AY286009.1 | - | 14 - 15 | 3649 | 89 | |
More Information
Set Membership: |
|

DML1
Genomic Map