Assay Details
| Gene Symbol: RPL24A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129217.5 | NP_565851.1 | 4 - 5 | 529 | 66 | |
| GenBank mRNA | AJ293729.1 | - | 362 | 66 | ||
| AY058086.1 | - | 4 - 5 | 373 | 66 | ||
| AY072350.1 | - | 4 - 5 | 375 | 66 | ||
| AY085323.1 | - | 4 - 5 | 381 | 66 | ||
| AY114728.1 | - | 3 - 4 | 335 | 66 | ||
More Information
Set Membership: |
|

RPL24A
Genomic Map