Assay Details
Gene Symbol: RAX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336612.1 | NP_001324751.1 | 2 - 3 | 472 | 61 | |
NM_129245.3 | NP_181226.1 | 2 - 3 | 389 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
