Assay Details
| Gene Symbol: PAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129260.3 | NP_181241.1 | 1 - 2 | 946 | 74 | |
| GenBank mRNA | AY045919.1 | - | 1 - 2 | 554 | 74 | |
| AY079363.1 | - | 1 - 2 | 428 | 74 | ||
| AY303128.1 | - | 1 - 2 | 428 | 74 | ||
| BT003330.1 | - | 1 - 2 | 554 | 74 | ||
More Information
Set Membership: |
|

PAL1
Genomic Map