Assay Details
| Gene Symbol: AT2G37110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129267.5 | NP_565858.1 | 1 - 2 | 428 | 61 | |
| GenBank mRNA | AK226303.1 | - | 1 - 2 | 372 | 61 | |
| BT024466.1 | - | 1 - 2 | 291 | 61 | ||
More Information
Set Membership: |
|

AT2G37110
Genomic Map