Assay Details
| Gene Symbol: AT2G37110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129267.5 | NP_565858.1 | 2 - 3 | 629 | 67 | |
| GenBank mRNA | AK226303.1 | - | 2 - 3 | 573 | 67 | |
| BT024466.1 | - | 2 - 3 | 492 | 67 | ||
More Information
Set Membership: |
|

AT2G37110
Genomic Map