Assay Details
Gene Symbol: ABCG33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336645.1 | NP_001325069.1 | 10 - 11 | 1782 | 124 | |
NM_001336646.1 | NP_001325068.1 | 9 - 10 | 1803 | 124 | ||
NM_001336647.1 | NP_001325070.1 | 9 - 10 | 1784 | 124 | ||
NM_129284.2 | NP_181265.1 | 10 - 11 | 1782 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
