Assay Details
Gene Symbol: AGD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036426.1 | NP_001031503.1 | 2 - 3 | 940 | 66 | |
NM_129310.3 | NP_181291.1 | 2 - 3 | 1010 | 66 | ||
GenBank mRNA | AB017876.1 | - | 2 - 3 | 1012 | 66 | |
AF436828.1 | - | 2 - 3 | 927 | 66 | ||
AY034983.1 | - | 2 - 3 | 926 | 66 | ||
AY063077.1 | - | 2 - 3 | 865 | 66 | ||
AY103310.1 | - | 2 - 3 | 865 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
