Assay Details
| Gene Symbol: AT2G37870 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129343.2 | NP_565872.1 | 1 - 2 | 404 | 86 | |
| GenBank mRNA | AY054635.1 | - | 1 - 2 | 369 | 86 | |
| AY072489.1 | - | 1 - 2 | 328 | 86 | ||
More Information
Set Membership: |
|

AT2G37870
Genomic Map