Assay Details
| Gene Symbol: AT2G38000 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129357.4 | NP_181336.1 | 2 - 3 | 337 | 70 | |
| GenBank mRNA | AY080880.1 | - | 2 - 3 | 317 | 70 | |
| AY114047.1 | - | 2 - 3 | 222 | 70 | ||
More Information
Set Membership: |
|

AT2G38000
Genomic Map