Assay Details
Gene Symbol: VCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084551.1 | NP_001078020.1 | 3 - 4 | 1239 | 93 | |
NM_129359.4 | NP_565879.1 | 3 - 4 | 1240 | 93 | ||
GenBank mRNA | AF359240.1 | - | 2 - 3 | 1147 | 93 | |
AY048248.1 | - | 3 - 4 | 1214 | 93 | ||
AY139799.1 | - | 2 - 3 | 1131 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
