Assay Details
Gene Symbol: VCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084551.1 | NP_001078020.1 | 9 - 10 | 1896 | 83 | |
NM_129359.4 | NP_565879.1 | 9 - 10 | 1897 | 83 | ||
GenBank mRNA | AF359240.1 | - | 8 - 9 | 1804 | 83 | |
AY048248.1 | - | 9 - 10 | 1871 | 83 | ||
AY139799.1 | - | 8 - 9 | 1788 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
