Assay Details
| Gene Symbol: AT2G38480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129405.4 | NP_565891.1 | 1 - 2 | 460 | 142 | |
| GenBank mRNA | AF370348.1 | - | 1 - 2 | 345 | 142 | |
| AY062978.1 | - | 1 - 2 | 243 | 142 | ||
More Information
Set Membership: |
|

AT2G38480
Genomic Map