Assay Details
| Gene Symbol: AT2G38480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129405.4 | NP_565891.1 | 2 - 3 | 596 | 75 | |
| GenBank mRNA | AF370348.1 | - | 2 - 3 | 481 | 75 | |
| AY062978.1 | - | 2 - 3 | 379 | 75 | ||
More Information
Set Membership: |
|

AT2G38480
Genomic Map