Assay Details
| Gene Symbol: GAUT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202773.2 | NP_001189702.1 | 9 - 10 | 1861 | 96 | |
| NM_129422.4 | NP_565893.1 | 9 - 10 | 1941 | 96 | ||
| GenBank mRNA | AY050982.1 | - | 9 - 10 | 1785 | 96 | |
| AY091448.1 | - | 9 - 10 | 1588 | 96 | ||
| KJ138859.1 | - | 9 - 10 | 1588 | 96 | ||
More Information
Set Membership: |
|

GAUT7
Genomic Map