Assay Details
| Gene Symbol: PIP2E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336755.1 | NP_001324169.1 | 3 - 4 | 1659 | 75 | |
| NM_129458.3 | NP_181434.1 | 3 - 4 | 1020 | 75 | ||
| GenBank mRNA | AY045690.1 | - | 3 - 4 | 820 | 75 | |
| AY054142.1 | - | 3 - 4 | 736 | 75 | ||
| AY057559.1 | - | 3 - 4 | 818 | 75 | ||
More Information
Set Membership: |
|

PIP2E
Genomic Map