Assay Details
| Gene Symbol: AT2G39690 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336785.1 | NP_001318384.1 | 7 - 8 | 1208 | 63 | |
| NM_001336790.1 | NP_001324846.1 | 7 - 8 | 1208 | 63 | ||
| NM_001336791.1 | NP_001324849.1 | 6 - 7 | 1273 | 63 | ||
More Information
Set Membership: |
|

AT2G39690
Genomic Map