Assay Details
Gene Symbol: ABI4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129580.2 | NP_181551.1 | 1 | 911 | 87 | |
GenBank mRNA | DQ446612.1 | - | 1 | 911 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
