Assay Details
Gene Symbol: UBP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336876.1 | NP_001323724.1 | 11 - 12 | 2636 | 67 | |
NM_129656.4 | NP_565944.1 | 11 - 12 | 2695 | 67 | ||
GenBank mRNA | AF048705.1 | - | 11 - 12 | 2600 | 67 | |
AY099701.1 | - | 11 - 12 | 2591 | 67 | ||
BT000300.1 | - | 11 - 12 | 2359 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
