Assay Details
Gene Symbol: UBP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336876.1 | NP_001323724.1 | 12 - 13 | 2919 | 104 | |
NM_129656.4 | NP_565944.1 | 12 - 13 | 2978 | 104 | ||
GenBank mRNA | AF048705.1 | - | 12 - 13 | 2883 | 104 | |
AY099701.1 | - | 12 - 13 | 2874 | 104 | ||
BT000300.1 | - | 12 - 13 | 2642 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
