Assay Details
| Gene Symbol: UBP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336876.1 | NP_001323724.1 | 2 - 3 | 673 | 62 | |
| NM_129656.4 | NP_565944.1 | 2 - 3 | 732 | 62 | ||
| GenBank mRNA | AF048705.1 | - | 2 - 3 | 637 | 62 | |
| AY099701.1 | - | 2 - 3 | 628 | 62 | ||
| BT000300.1 | - | 2 - 3 | 396 | 62 | ||
More Information
Set Membership: |
|

UBP5
Genomic Map