Assay Details
| Gene Symbol: UBP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336876.1 | NP_001323724.1 | 4 - 5 | 879 | 118 | |
| NM_129656.4 | NP_565944.1 | 4 - 5 | 938 | 118 | ||
| GenBank mRNA | AF048705.1 | - | 4 - 5 | 843 | 118 | |
| AY099701.1 | - | 4 - 5 | 834 | 118 | ||
| BT000300.1 | - | 4 - 5 | 602 | 118 | ||
More Information
Set Membership: |
|

UBP5
Genomic Map