Assay Details
Gene Symbol: UBP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336876.1 | NP_001323724.1 | 9 - 10 | 1930 | 122 | |
NM_129656.4 | NP_565944.1 | 9 - 10 | 1989 | 122 | ||
GenBank mRNA | AF048705.1 | - | 9 - 10 | 1894 | 122 | |
AY099701.1 | - | 9 - 10 | 1885 | 122 | ||
BT000300.1 | - | 9 - 10 | 1653 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
