Assay Details
| Gene Symbol: CRK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129679.2 | NP_181647.1 | 10 - 11 | 1992 | 61 | |
| GenBank mRNA | AF153351.1 | - | 10 - 11 | 1700 | 61 | |
| AF435448.1 | - | 10 - 11 | 1700 | 61 | ||
More Information
Set Membership: |
|

CRK1
Genomic Map