Assay Details
| Gene Symbol: CRK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129679.2 | NP_181647.1 | 7 - 8 | 1648 | 138 | |
| GenBank mRNA | AF153351.1 | - | 7 - 8 | 1356 | 138 | |
| AF435448.1 | - | 7 - 8 | 1356 | 138 | ||
More Information
Set Membership: |
|

CRK1
Genomic Map