Assay Details
| Gene Symbol: AT2G41620 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129725.5 | NP_565951.1 | 13 - 14 | 2325 | 79 | |
| GenBank mRNA | AY034907.1 | - | 13 - 14 | 2270 | 79 | |
| AY113905.1 | - | 13 - 14 | 2219 | 79 | ||
More Information
Set Membership: |
|

AT2G41620
Genomic Map