Assay Details
Gene Symbol: GK-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336938.1 | NP_001323627.1 | 9 - 10 | 1483 | 116 | |
NM_001336939.1 | NP_001323629.1 | 8 - 9 | 1561 | 116 | ||
NM_001336940.1 | NP_001323628.1 | 9 - 10 | 1487 | 116 | ||
NM_001336941.1 | NP_001323630.1 | 9 - 10 | 1478 | 116 | ||
NM_001336943.1 | NP_001323632.1 | 10 - 11 | 1352 | 116 | ||
NM_001336945.1 | NP_001323634.1 | 9 - 10 | 1299 | 116 | ||
NM_129752.5 | NP_565961.1 | 10 - 11 | 1409 | 116 | ||
GenBank mRNA | BT028912.1 | - | 8 - 9 | 1072 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
