Assay Details
Gene Symbol: GK-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336940.1 | NP_001323628.1 | 8 - 9 | 1276 | 100 | |
NM_001336941.1 | NP_001323630.1 | 7 - 8 | 1346 | 100 | ||
NM_001336942.1 | NP_001323631.1 | 8 - 9 | 1233 | 100 | ||
NM_001336943.1 | NP_001323632.1 | 8 - 9 | 1220 | 100 | ||
NM_001336944.1 | NP_001323633.1 | 8 - 9 | 1219 | 100 | ||
NM_129752.5 | NP_565961.1 | 8 - 9 | 1277 | 100 | ||
GenBank mRNA | BT028912.1 | - | 6 - 7 | 940 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
