Assay Details
Gene Symbol: GK-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336938.1 | NP_001323627.1 | 8 - 9 | 1403 | 157 | |
NM_001336941.1 | NP_001323630.1 | 8 - 9 | 1398 | 157 | ||
NM_001336942.1 | NP_001323631.1 | 9 - 10 | 1285 | 252 | ||
NM_001336943.1 | NP_001323632.1 | 9 - 10 | 1272 | 157 | ||
NM_001336944.1 | NP_001323633.1 | 9 - 10 | 1271 | 172 | ||
NM_129752.5 | NP_565961.1 | 9 - 10 | 1329 | 157 | ||
GenBank mRNA | BT028912.1 | - | 7 - 8 | 992 | 157 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
