Assay Details
| Gene Symbol: AT2G42080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129771.4 | NP_181738.1 | 3 - 4 | 530 | 99 | |
| GenBank mRNA | AY136439.1 | - | 3 - 4 | 344 | 99 | |
| BT002128.1 | - | 3 - 4 | 332 | 99 | ||
More Information
Set Membership: |
|

AT2G42080
Genomic Map