Assay Details
| Gene Symbol: TTL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129819.3 | NP_565976.1 | 6 - 7 | 2302 | 103 | |
| GenBank mRNA | AF367321.1 | - | 6 - 7 | 2066 | 103 | |
| AY143941.1 | - | 6 - 7 | 1926 | 103 | ||
More Information
Set Membership: |
|

TTL3
Genomic Map