Assay Details
Gene Symbol: MSS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129891.2 | NP_565996.1 | 1 | 761 | 150 | |
GenBank mRNA | AF250344.1 | - | 1 | 715 | 150 | |
AY062815.1 | - | 1 | 759 | 150 | ||
AY081574.1 | - | 1 | 669 | 150 | ||
AY087087.1 | - | 1 | 761 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
