Assay Details
Gene Symbol: GLX2-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337029.1 | NP_001323646.1 | 2 - 3 | 343 | 84 | |
NM_129904.3 | NP_565999.1 | 2 - 3 | 519 | 84 | ||
NM_201950.3 | NP_973679.1 | 1 - 2 | 899 | 84 | ||
GenBank mRNA | AY063806.1 | - | 2 - 3 | 404 | 84 | |
AY091278.1 | - | 2 - 3 | 249 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
