Assay Details
Gene Symbol: MPK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129941.4 | NP_181907.1 | 2 - 3 | 596 | 71 | |
GenBank mRNA | AY120737.1 | - | 2 - 3 | 479 | 71 | |
BT008855.1 | - | 2 - 3 | 365 | 71 | ||
D21842.1 | - | 2 - 3 | 437 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
