Assay Details
Gene Symbol: MPK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129941.4 | NP_181907.1 | 3 - 4 | 729 | 136 | |
GenBank mRNA | AY120737.1 | - | 3 - 4 | 612 | 136 | |
BT008855.1 | - | 3 - 4 | 498 | 136 | ||
D21842.1 | - | 3 - 4 | 570 | 136 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
