Assay Details
Gene Symbol: MPK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129941.4 | NP_181907.1 | 5 - 6 | 1238 | 100 | |
GenBank mRNA | AY120737.1 | - | 5 - 6 | 1121 | 100 | |
BT008855.1 | - | 5 - 6 | 1007 | 100 | ||
D21842.1 | - | 5 - 6 | 1079 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
