Assay Details
| Gene Symbol: MLO15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129974.1 | NP_181939.1 | 10 - 11 | 923 | 98 | |
| NM_201957.2 | NP_973686.1 | 10 - 11 | 1069 | 98 | ||
| GenBank mRNA | AF369576.1 | - | 10 - 11 | 923 | 98 | |
More Information
Set Membership: |
|

MLO15
Genomic Map