Assay Details
Gene Symbol: MLO15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129974.1 | NP_181939.1 | 12 - 13 | 1196 | 98 | |
NM_201957.2 | NP_973686.1 | 12 - 13 | 1342 | 98 | ||
GenBank mRNA | AF369576.1 | - | 12 - 13 | 1196 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
