Assay Details
| Gene Symbol: ASHH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129978.4 | NP_566010.1 | 12 - 13 | 1396 | 116 | |
| GenBank mRNA | AY045886.1 | - | 12 - 13 | 1283 | 116 | |
More Information
Set Membership: |
|

ASHH3
Genomic Map