Assay Details
| Gene Symbol: ASHH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129978.4 | NP_566010.1 | 7 - 8 | 993 | 87 | |
| GenBank mRNA | AY045886.1 | - | 7 - 8 | 880 | 87 | |
| AY091447.1 | - | 7 - 8 | 699 | 87 | ||
More Information
Set Membership: |
|

ASHH3
Genomic Map